Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gr28a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078780.5 | NP_523504.2 | 1 - 2 | 672 | 78 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Recommended Assay Drosophila melanogaster |