Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: robl37BC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136087.3 | NP_609931.1 | 1 | 439 | 75 | |
GenBank mRNA | BT023356.1 | - | 1 | 353 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |