Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136090.3 | NP_609934.3 | 2 - 3 | 943 | 132 | |
GenBank mRNA | AY071630.1 | - | 2 - 3 | 768 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |