Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phlpp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273629.1 | NP_001260558.1 | 4 - 5 | 2289 | 67 | |
NM_136094.2 | NP_609938.1 | 4 - 5 | 2104 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |