Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: p47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136440.6 | NP_610284.1 | 3 - 4 | 829 | 78 | |
GenBank mRNA | AY058260.1 | - | 3 - 4 | 801 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |