Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nito | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136495.3 | NP_610339.1 | 4 - 5 | 2902 | 89 | |
GenBank mRNA | AF145664.1 | - | 4 - 5 | 2879 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |