Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkc53E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103880.3 | NP_001097350.1 | 14 - 15 | 2262 | 76 | |
NM_001259448.1 | NP_001246377.1 | 9 - 10 | 1593 | 76 | ||
NM_001259449.1 | NP_001246378.1 | 14 - 15 | 2202 | 76 | ||
NM_001274106.1 | NP_001261035.1 | 13 - 14 | 2261 | 76 | ||
NM_057334.3 | NP_476682.1 | 13 - 14 | 2744 | 76 | ||
NM_166201.3 | NP_725626.1 | 14 - 15 | 2265 | 76 | ||
GenBank mRNA | AY095003.1 | - | 14 - 15 | 2236 | 76 | |
BT120171.1 | - | 9 - 10 | 1593 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |