Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fat-spondin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058087.5 | NP_477435.1 | 4 - 5 | 1368 | 91 | |
GenBank mRNA | AF135119.1 | - | 4 - 5 | 1341 | 91 | |
BT031193.1 | - | 4 - 5 | 1331 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |