Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cora | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079067.4 | NP_523791.2 | 12 - 13 | 5034 | 81 | |
NM_166338.3 | NP_725866.1 | 11 - 12 | 2037 | 81 | ||
GenBank mRNA | BT133208.1 | - | 11 - 12 | 2035 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |