Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hts | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103907.3 | NP_001097377.2 | 7 - 8 | 1769 | 77 | |
NM_001169734.2 | NP_001163205.1 | 7 - 8 | 1759 | 77 | ||
NM_001169735.2 | NP_001163206.1 | 6 - 7 | 2005 | 77 | ||
NM_001202048.3 | NP_001188977.2 | 5 - 6 | 1516 | 77 | ||
NM_001259491.2 | NP_001246420.1 | 7 - 8 | 1769 | 77 | ||
NM_001259492.2 | NP_001246421.1 | 6 - 7 | 1565 | 77 | ||
NM_001299683.1 | NP_001286612.1 | 5 - 6 | 1571 | 77 | ||
NM_001382109.1 | NP_001369098.1 | 7 - 8 | 1769 | 77 | ||
NM_057529.4 | NP_476877.1 | 7 - 8 | 1769 | 77 | ||
NM_166347.3 | NP_725886.2 | 7 - 8 | 1769 | 77 | ||
NM_166348.3 | NP_725887.2 | 7 - 8 | 1759 | 77 | ||
NM_166349.3 | NP_725888.2 | 6 - 7 | 2005 | 77 | ||
NM_166350.2 | NP_725889.1 | 7 - 8 | 1769 | 77 | ||
GenBank mRNA | AF151705.1 | - | 7 - 8 | 1731 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |