Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fak | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144246.3 | NP_001137718.1 | 13 - 14 | 4599 | 82 | |
NM_001144247.3 | NP_001137719.2 | 14 - 15 | 3128 | 82 | ||
NM_001169736.2 | NP_001163207.1 | 13 - 14 | 3693 | 82 | ||
NM_079069.4 | NP_523793.2 | 13 - 14 | 3699 | 82 | ||
NM_166352.3 | NP_725891.1 | 14 - 15 | 3485 | 82 | ||
NM_166353.3 | NP_725892.1 | 14 - 15 | 3226 | 82 | ||
GenBank mRNA | BT133413.1 | - | 13 - 14 | 3682 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |