Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fak | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144246.3 | NP_001137718.1 | 7 - 8 | 2529 | 68 | |
NM_001144247.3 | NP_001137719.2 | 8 - 9 | 1958 | 68 | ||
NM_001169736.2 | NP_001163207.1 | 7 - 8 | 2529 | 68 | ||
NM_079069.4 | NP_523793.2 | 7 - 8 | 2529 | 68 | ||
NM_166352.3 | NP_725891.1 | 8 - 9 | 2315 | 68 | ||
NM_166353.3 | NP_725892.1 | 8 - 9 | 2056 | 68 | ||
GenBank mRNA | BT133413.1 | - | 7 - 8 | 2512 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |