Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hil | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137649.3 | NP_611493.2 | 7 - 8 | 2456 | 60 | |
NM_166402.2 | NP_725980.1 | 7 - 8 | 2519 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |