Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fem-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137664.3 | NP_611508.1 | 3 - 4 | 1409 | 76 | |
NM_166408.2 | NP_725993.1 | 4 - 5 | 1216 | 76 | ||
GenBank mRNA | BT133040.1 | - | 3 - 4 | 1346 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |