Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: exu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169747.2 | NP_001163218.1 | 4 - 5 | 1766 | 82 | |
NM_166412.3 | NP_725997.1 | 3 - 4 | 1756 | 82 | ||
GenBank mRNA | BT126187.1 | - | 4 - 5 | 1780 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |