Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dom | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299747.1 | NP_001286676.1 | 12 - 13 | 6326 | 60 | |
NM_080094.4 | NP_524833.2 | 11 - 12 | 6221 | 60 | ||
NM_166446.3 | NP_726065.1 | 11 - 12 | 6221 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |