Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nahoda | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259545.2 | NP_001246474.1 | 9 - 10 | 4086 | 66 | |
NM_079091.4 | NP_523815.2 | 9 - 10 | 4086 | 66 | ||
NM_166569.3 | NP_726264.1 | 9 - 10 | 3618 | 66 | ||
GenBank mRNA | AY060632.1 | - | 9 - 10 | 4043 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |