Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4e3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078803.2 | NP_523527.1 | 2 - 3 | 315 | 94 | |
GenBank mRNA | BT030822.1 | - | 2 - 3 | 315 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |