Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032069.2 | NP_001027240.1 | 10 - 11 | 2148 | 83 | |
NM_001032072.2 | NP_001027243.2 | 8 - 9 | 1452 | 83 | ||
NM_078817.4 | NP_523541.2 | 10 - 11 | 2148 | 83 | ||
GenBank mRNA | BT010016.1 | - | 10 - 11 | 2147 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |