Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032069.2 | NP_001027240.1 | 13 - 14 | 2780 | 71 | |
NM_001032072.2 | NP_001027243.2 | 12 - 13 | 1975 | 71 | ||
NM_078817.4 | NP_523541.2 | 14 - 15 | 2671 | 71 | ||
GenBank mRNA | BT010016.1 | - | 14 - 15 | 2670 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |