Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsp42Ee | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273824.1 | NP_001260753.1 | 4 - 5 | 709 | 57 | |
NM_078907.3 | NP_523631.1 | 4 - 5 | 722 | 57 | ||
GenBank mRNA | AF220043.1 | - | 4 - 5 | 690 | 57 | |
BT044107.1 | - | 4 - 5 | 720 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |