Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tou | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103800.3 | NP_001097270.1 | 7 - 8 | 3527 | 88 | |
NM_001273969.1 | NP_001260898.1 | 8 - 9 | 3054 | 88 | ||
NM_001273970.1 | NP_001260899.1 | 8 - 9 | 3493 | 88 | ||
NM_078977.5 | NP_523701.3 | 7 - 8 | 3527 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |