Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gbeta76C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078996.3 | NP_523720.2 | 2 - 3 | 609 | 66 | |
GenBank mRNA | AY118886.1 | - | 2 - 3 | 580 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |