Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078999.4 | NP_523723.1 | 7 - 8 | 2757 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |