Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136511.3 | NP_610355.1 | 2 - 3 | 1457 | 86 | |
GenBank mRNA | AY070546.1 | - | 2 - 3 | 1468 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |