Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: coil | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014505.2 | NP_001014505.1 | 4 - 5 | 1799 | 64 | |
NM_001299255.1 | NP_001286184.1 | 4 - 5 | 1796 | 64 | ||
GenBank mRNA | BT050478.1 | - | 4 - 5 | 1799 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |