Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rme-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299307.1 | NP_001286236.1 | 10 - 11 | 2497 | 76 | |
NM_136623.2 | NP_610467.1 | 10 - 11 | 2500 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |