Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4p3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136629.3 | NP_610473.1 | 2 - 3 | 438 | 118 | |
GenBank mRNA | AY075201.1 | - | 2 - 3 | 417 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |