Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136633.4 | NP_610477.2 | 1 - 2 | 233 | 73 | |
GenBank mRNA | AF123262.1 | - | 1 - 2 | 179 | 73 | |
BT044526.1 | - | 1 - 2 | 223 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |