Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vamp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299326.1 | NP_001286255.1 | 3 - 4 | 815 | 97 | |
NM_136680.4 | NP_610524.1 | 3 - 4 | 757 | 97 | ||
GenBank mRNA | AY071695.1 | - | 3 - 4 | 769 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |