Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ir48c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136856.1 | NP_610700.1 | 1 | 540 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |