Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: jeb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273981.1 | NP_001260910.1 | 2 - 3 | 2902 | 62 | |
NM_136882.3 | NP_610726.1 | 2 - 3 | 2415 | 62 | ||
GenBank mRNA | AF425733.1 | - | 2 - 3 | 2416 | 62 | |
AF425734.1 | - | 3 - 4 | 3756 | 62 | ||
AY060310.1 | - | 2 - 3 | 2434 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |