Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169803.2 | NP_001163274.1 | 18 - 19 | 2995 | 70 | |
NM_001169804.2 | NP_001163275.2 | 14 - 15 | 3155 | 70 | ||
NM_001169805.2 | NP_001163276.1 | 17 - 18 | 2822 | 70 | ||
NM_001169807.2 | NP_001163278.2 | 17 - 18 | 3108 | 70 | ||
NM_137970.5 | NP_611814.3 | 18 - 19 | 2826 | 70 | ||
NM_166616.3 | NP_726350.1 | 17 - 18 | 3108 | 70 | ||
NM_166617.2 | NP_726351.1 | 15 - 16 | 2242 | 70 | ||
NM_166618.3 | NP_726352.1 | 16 - 17 | 2591 | 70 | ||
NM_166619.2 | NP_726353.1 | 5 - 6 | 1115 | 70 | ||
GenBank mRNA | AY060327.1 | - | 5 - 6 | 1115 | 70 | |
BT021326.1 | - | 18 - 19 | 2822 | 70 | ||
BT133111.1 | - | 16 - 17 | 2591 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |