Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pio | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169824.2 | NP_001163295.1 | 6 - 7 | 1719 | 95 | |
NM_001299920.1 | NP_001286849.1 | 6 - 7 | 965 | 95 | ||
NM_166677.3 | NP_726461.1 | 7 - 8 | 1247 | 95 | ||
GenBank mRNA | AY862157.1 | - | 7 - 8 | 1217 | 95 | |
BT003795.1 | - | 6 - 7 | 1702 | 95 | ||
BT011382.1 | - | 7 - 8 | 1215 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |