Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eps-15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259573.1 | NP_001246502.1 | 8 - 9 | 2602 | 82 | |
NM_138121.2 | NP_611965.2 | 9 - 10 | 2665 | 82 | ||
NM_166686.1 | NP_726481.1 | 8 - 9 | 2602 | 82 | ||
NM_166687.1 | NP_726482.1 | 8 - 9 | 2602 | 82 | ||
GenBank mRNA | BT125713.1 | - | 8 - 9 | 2568 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |