Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258482.2 | NP_001245411.1 | 3 - 4 | 567 | 121 | |
NM_001258483.2 | NP_001245412.1 | 3 - 4 | 591 | 121 | ||
NM_143651.6 | NP_651908.1 | 3 - 4 | 591 | 121 | ||
NM_166743.3 | NP_726549.1 | 4 - 5 | 516 | 121 | ||
NM_166744.3 | NP_726550.1 | 3 - 4 | 537 | 121 | ||
GenBank mRNA | AF112976.1 | - | 3 - 4 | 501 | 121 | |
AY061094.1 | - | 3 - 4 | 419 | 121 | ||
BT012456.1 | - | 3 - 4 | 589 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |