Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zyx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258501.3 | NP_001245430.1 | 3 - 4 | 739 | 84 | |
NM_143758.4 | NP_652015.1 | 3 - 4 | 842 | 84 | ||
NM_166815.3 | NP_726638.1 | 2 - 3 | 569 | 84 | ||
NM_166819.3 | NP_726642.1 | 4 - 5 | 961 | 84 | ||
GenBank mRNA | AF219948.1 | - | 3 - 4 | 795 | 84 | |
BT150084.1 | - | 2 - 3 | 538 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |