Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mnt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169181.3 | NP_001162652.1 | 2 - 3 | 1009 | 63 | |
NM_001258584.2 | NP_001245513.1 | 2 - 3 | 1099 | 63 | ||
NM_166983.3 | NP_726879.1 | 2 - 3 | 1099 | 63 | ||
NM_166984.3 | NP_726880.1 | 2 - 3 | 1240 | 63 | ||
GenBank mRNA | AY075354.1 | - | 1 - 2 | 989 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |