Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RpI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079019.3 | NP_523743.1 | 6 - 7 | 3882 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |