Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fidipidine | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079036.5 | NP_523760.1 | 6 - 7 | 1318 | 65 | |
GenBank mRNA | BT031141.1 | - | 6 - 7 | 1250 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |