Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acox57D-d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079079.4 | NP_523803.1 | 4 - 5 | 1101 | 64 | |
GenBank mRNA | AY070539.1 | - | 4 - 5 | 1110 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |