Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gr59b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079094.3 | NP_523818.2 | 1 - 2 | 948 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |