Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079214.4 | NP_523938.2 | 2 - 3 | 813 | 62 | |
GenBank mRNA | AY089550.1 | - | 3699 | 62 | ||
BT089013.1 | - | 2 - 3 | 670 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |