Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: melt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014572.4 | NP_001014572.1 | 3 - 4 | 1116 | 77 | |
NM_001274566.1 | NP_001261495.1 | 4 - 5 | 1402 | 77 | ||
NM_079229.4 | NP_523953.2 | 3 - 4 | 1116 | 77 | ||
GenBank mRNA | BT029129.1 | - | 3 - 4 | 1090 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |