Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: foi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274655.1 | NP_001261584.1 | 4 - 5 | 1173 | 82 | |
NM_001274656.1 | NP_001261585.1 | 4 - 5 | 1250 | 82 | ||
NM_001300056.1 | NP_001286985.1 | 4 - 5 | 1565 | 82 | ||
NM_079250.2 | NP_523974.3 | 4 - 5 | 1565 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |