Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pi3K68D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169949.2 | NP_001163420.1 | 10 - 11 | 4142 | 109 | |
NM_079304.4 | NP_524028.2 | 9 - 10 | 4491 | 109 | ||
NM_168475.3 | NP_729743.1 | 10 - 11 | 4190 | 109 | ||
GenBank mRNA | AY058587.1 | - | 9 - 10 | 4433 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |