Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ara | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169961.2 | NP_001163432.1 | 2 - 3 | 1031 | 62 | |
NM_079321.3 | NP_524045.2 | 2 - 3 | 1031 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |