Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hml | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274880.1 | NP_001261809.1 | 22 - 23 | 10630 | 112 | |
NM_079336.3 | NP_524060.2 | 22 - 23 | 10633 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |