Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298569.1 | NP_001285498.1 | 2 - 3 | 1008 | 83 | |
NM_079356.3 | NP_524080.1 | 7 - 8 | 2749 | 83 | ||
GenBank mRNA | AF199466.1 | - | 7 - 8 | 2703 | 83 | |
AY051953.1 | - | 7 - 8 | 2722 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |