Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SsRbeta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079379.3 | NP_524103.2 | 2 - 3 | 477 | 100 | |
GenBank mRNA | AF160923.1 | - | 2 - 3 | 454 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |